Learn More
Sigma Aldrich Fine Chemicals Biosciences Complete Whole Transcripto
Catalog No. 502914680
Encompass
WTA2 is optimized to amplify RNA from formalin-fixed paraffin-embedded (ffPE) and other damaged or degraded samples. Whole Transcriptome Amplification (WTA) technology allows for representative amplification of low nanogram quantities of total RNA in less than 4 hours without 3-bias. Amplification products are suitable for applications such as qPCR micro array analysis and cloning. The WTA2 kit contains the polymerase needed to amplify the cDNA library.
Catalog No. 50-291-4680 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. WTA250RXN
WTA2 is optimized to amplify RNA from formalin-fixed paraffin-embedded (ffPE) and other damaged or degraded samples. Whole Transcriptome Amplification (WTA) technology allows for representative amplification of low nanogram quantities of total RNA in less than 4 hours without 3-bias. Amplification products are suitable for applications such as qPCR micro array analysis and cloning. The WTA2 kit contains the polymerase needed to amplify the cDNA library.