Learn More
TRIBIOSCIENCE INC Mitochondrial Complex I Activity Colorimetric Assay, 100 Tests
SDP
Catalog No. NC3454568
Encompass_Preferred
Tribioscience’s Mitochondrial Complex I Activity Colorimetric Assay kit uses decylubiquinone, an analog of ubiquinone, as an electron acceptor that gets converted to decylubiquinol through the catalytic activity of Complex I. Complex I activity is determined by recording the change in absorbance (A600) of the reduced Complex I dye. Specific Complex I activity is obtained by subtracting the activity in presence of Complex I inhibitor rotenone from total activity. This kit can detect as low as 0.1 mU/well and is linear up to 7 mU/well.
Catalog No. NC3454568 Supplier TRIBIOSCIENCE INC Supplier No. TBS2017
Tribioscience’s Mitochondrial Complex I Activity Colorimetric Assay kit uses decylubiquinone, an analog of ubiquinone, as an electron acceptor that gets converted to decylubiquinol through the catalytic activity of Complex I. Complex I activity is determined by recording the change in absorbance (A600) of the reduced Complex I dye. Specific Complex I activity is obtained by subtracting the activity in presence of Complex I inhibitor rotenone from total activity. This kit can detect as low as 0.1 mU/well and is linear up to 7 mU/well.