Learn More
Abcam Complex I Enzyme Activity Dipstick Assay Kit, 48TEST
Catalog No. 502585054
Encompass_Preferred
Complex I Enzyme Activity Dipstick Assay Kit is an immunoassay kit for the detection of Complex I Enzyme Activity in Mouse, Rat, Cow, Human in Cell culture extracts, Tissue, Cell culture supernatant samples.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-258-5054 Supplier Abcam Supplier No. AB10972048TEST
Complex I Enzyme Activity Dipstick Assay Kit is an immunoassay kit for the detection of Complex I Enzyme Activity in Mouse, Rat, Cow, Human in Cell culture extracts, Tissue, Cell culture supernatant samples.
The product is subject to the following: Abcam Restricted Use Statement