Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Complex III Subunit 5 antibody (YA1865) | 10 UL
SDP

Catalog No. 5000441505
Encompass_Preferred

Complex III Subunit 5 antibody (YA1865) is a Rabbit-derived, non-conjugated IgG monoclonal antibody targeting Complex III Subunit 5. This subunit is a component of the ubiquinol-cytochrome c oxidoreductase, also known as cytochrome b-c1 complex or complex III (CIII), which is a multisubunit transmembrane complex involved in the mitochondrial electron transport chain. It catalyzes electron transfer from ubiquinol to cytochrome c, coupling this redox reaction to proton translocation across the mitochondrial inner membrane.

  • Rabbit-derived, non-conjugated IgG monoclonal antibody
  • Targets Complex III Subunit 5
  • Recombinant, monoclonal
  • Species reactivity: Human, mouse, rat
  • Immunogen: Synthetic peptide of human RISP
  • Multiple applications: WB, IHC-P, IHC-F, ICC/IF, IP
  • Sensitivity: Endogenous
  • Isotype: IgG

Catalog No. 50-004-41505 Supplier Medchemexpress LLC Supplier No. HYP8212010UL
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Complex III Subunit 5 antibody (YA1865) is a Rabbit-derived, non-conjugated IgG monoclonal antibody targeting Complex III Subunit 5. This subunit is a component of the ubiquinol-cytochrome c oxidoreductase, also known as cytochrome b-c1 complex or complex III (CIII), which is a multisubunit transmembrane complex involved in the mitochondrial electron transport chain. It catalyzes electron transfer from ubiquinol to cytochrome c, coupling this redox reaction to proton translocation across the mitochondrial inner membrane.

  • Rabbit-derived, non-conjugated IgG monoclonal antibody
  • Targets Complex III Subunit 5
  • Recombinant, monoclonal
  • Species reactivity: Human, mouse, rat
  • Immunogen: Synthetic peptide of human RISP
  • Multiple applications: WB, IHC-P, IHC-F, ICC/IF, IP
  • Sensitivity: Endogenous
  • Isotype: IgG

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.