Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Complex III Subunit 5 Antibody (YA1865) | 100 UL
SDP

Catalog No. 5000441310
Encompass_Preferred

Complex III Subunit 5 Antibody (YA1865) is a Rabbit-derived, non-conjugated IgG monoclonal antibody targeting Complex III Subunit 5. Intended for research use only, this antibody supports studies of ubiquinol-cytochrome c oxidoreductase (Complex III) in the mitochondrial electron transport chain.

  • Host: Rabbit
  • Clonality: Recombinant, monoclonal
  • Species reactivity: Human, mouse, rat
  • Applications: WB, IHC-P, IHC-F, ICC/IF, IP
  • Sensitivity: Endogenous
  • Conjugation: Non-conjugated
  • Isotype: IgG
  • Appearance: Liquid
  • Formulation: Supplied in Tris-Glycine, NaCl, Glycerol, Sodium azide, and BSA
  • Storage and stability: Stored at -20°C for 1 year. Avoid repeated freeze/thaw cycles.

Catalog No. 50-004-41310 Supplier Medchemexpress LLC Supplier No. HYP82120100UL
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Complex III Subunit 5 Antibody (YA1865) is a Rabbit-derived, non-conjugated IgG monoclonal antibody targeting Complex III Subunit 5. Intended for research use only, this antibody supports studies of ubiquinol-cytochrome c oxidoreductase (Complex III) in the mitochondrial electron transport chain.

  • Host: Rabbit
  • Clonality: Recombinant, monoclonal
  • Species reactivity: Human, mouse, rat
  • Applications: WB, IHC-P, IHC-F, ICC/IF, IP
  • Sensitivity: Endogenous
  • Conjugation: Non-conjugated
  • Isotype: IgG
  • Appearance: Liquid
  • Formulation: Supplied in Tris-Glycine, NaCl, Glycerol, Sodium azide, and BSA
  • Storage and stability: Stored at -20°C for 1 year. Avoid repeated freeze/thaw cycles.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.