Learn More
ERA WATERS CORP COMPLEX NUTRIENT 500ML
SDP
Catalog No. NC9076506
Encompass
One 500 mL whole-volume bottle is ready to analyze,maximum holding time of 2 months
Catalog No. NC9076506 Supplier ERA WATERS CORP Supplier No. 741
One 500 mL whole-volume bottle is ready to analyze,maximum holding time of 2 months