Learn More
MSC Honeywell Full Facepiece Respirator, Silicone, 5-Point Harness, Medium, 1 ea
Catalog No. 502370319
Encompass
The RU6500 Series is a silicone full facepiece designed for all-day comfort and affordability. It features a soft and flexible silicone facepiece seal for comfortable wear over extended periods. The mask has a lightweight design to reduce head and neck stress. Its polycarbonate lens is built to withstand high impact, eliminating the need for additional eye protection.
  • Soft, comfortable silicone nose cup for all-day wear
  • High-impact polycarbonate lens meets ANSI Z87, provides wide, clear field of vision
  • Five-point strap with center strap for secure, adjustable fit
  • Industrial mesh headnet option for comfort, usable under hard hat
  • Threaded cartridge connector prevents accidental disconnections
  • Adaptable to side/front mount CF-SAR and PAPR systems
Catalog No. 50-237-0319 Supplier MSC Supplier No. 69541340
The RU6500 Series is a silicone full facepiece designed for all-day comfort and affordability. It features a soft and flexible silicone facepiece seal for comfortable wear over extended periods. The mask has a lightweight design to reduce head and neck stress. Its polycarbonate lens is built to withstand high impact, eliminating the need for additional eye protection.
  • Soft, comfortable silicone nose cup for all-day wear
  • High-impact polycarbonate lens meets ANSI Z87, provides wide, clear field of vision
  • Five-point strap with center strap for secure, adjustable fit
  • Industrial mesh headnet option for comfort, usable under hard hat
  • Threaded cartridge connector prevents accidental disconnections
  • Adaptable to side/front mount CF-SAR and PAPR systems