Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Germaine™ Laboratories Compliance™ Gold iFOB

Catalog No. 23111309
Change view
Click to view available options
Quantity:
30 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-111-309 30 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 23-111-309 Supplier Germaine™ Laboratories Supplier No. 740302
 Restricted Item
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Germaine™ Laboratories Compliance Gold™ iFOB is a rapid chromatographic assay for the qualitative detection of fecal occult blood in human stool samples by laboratories or physician offices

  • Useful aid in the diagnosis and therapy of gastrointestinal disorders
  • Patient-friendly for use in physicians' offices, labs, hospitals, clinics, or homes as test sample flows up through absorbent device
  • Labeled antibody-dye conjugate binds to hemoglobin in a specimen, forming an antibody-antigen complex
  • Results within five minutes
  • Positive results: one band in the control (C) region, one band in the test (T) region
  • Negative results: only one band in the control (C) region

Specifications

Content And Storage 15°C to 30°C
CPT Code 82274QW, G0328QW
CE Marker Yes
Clia Complexity Waived
Detectable Analytes Fecal Occult Blood
Sensitivity 50 ng hHB/mL
Disposable Single-use
For Use With (Application) iFOB
Quantity 30 Tests
Test Time 5 min.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.