Learn More
Cayman Chemical Complanatuside, Purity: ≥95%, CAS: 116183-66-5, 10 mg
SDP
Catalog No. 502940172
Encompass_Preferred
A flavonoid glucoside; inhibits NO production in LPS-stimulated RAW 264.7 cells (IC50 = 21.2 μM)
Catalog No. 50-294-0172 Supplier Cayman Chemical Supplier No. 2729510 MG
A flavonoid glucoside; inhibits NO production in LPS-stimulated RAW 264.7 cells (IC50 = 21.2 μM)