Learn More
Cayman Chemical ComplnatusIde 50mg
SDP
Catalog No. 502939238
Encompass_Preferred
A flavonoid glucoside; inhibits NO production in LPS-stimulated RAW 264.7 cells (IC50 = 21.2 μM)
Catalog No. 50-293-9238 Supplier Cayman Chemical Supplier No. 2729550 MG
A flavonoid glucoside; inhibits NO production in LPS-stimulated RAW 264.7 cells (IC50 = 21.2 μM)