Learn More
AdipoGen Compound 112254 (10 mg)
SDP
Supplier:  AdipoGen AGCR10155M010
Encompass_Preferred
Chemical. CAS: 949745-75-9. Formula: C26H35N3O3. MW: 437.6. Adiponectin receptor (AdipoR) agonist. Binds to AdipoR1 and AdipoR2 less potent than AdipoRon (Prod. No. AG-CR1-0154) http://www.adipogen.com/ag-cr1-0154/adiporon.html . Activates 5'-adenosine monophosphate-activated protein kinase (AMPK) in cultured mammalian cells. Improves diabetes, glucose and lipid metabolism and insulin sensitivity in cultured cells and in mice by AdipoR-dependent mechanisms.
Catalog No. 50-205-3855