Learn More
Enzo Life Sciences Compound 15e (5mg). CAS: 371943-05-4
SDP
Supplier:  Enzo Life Sciences ALX270455M005
Encompass_Preferred
Potent and specific inhibitor of phosphoinositide 3-kinasep110α isoform (IC50=0.0020µM). p110β isoform (IC50=0.016µM)p110γ isoform (IC50=0.66µM)C2β isoform (IC50=0.22µM). Inhibits proliferation of A375 melanoma cells (IC50=0.58µM). Inhibits proliferation of U87MG human glioblastoma cells (IC50=1.10.µM).Inhibits proliferation of A2780 human ovarian cells (IC50=0.27µM). Alternative name: 3-[4-(4-Morpholinyl)thieno[3,2-d]pyrimidin-2-yl]-phenol. Purity: ≥99% (HPLC). Appearance: White to off-white solid. Solubility: Soluble in DMSO or dioxane. Long Term Storage: -20°C.
Catalog No. 50-200-8580