Learn More
AdipoGen Compound 34 (1 mg)
SDP
Supplier:  AdipoGen AGCR10007M001
Encompass_Preferred
Chemical. CAS: 564462-36-8. Formula: C31H24F3N3O3. MW: 543.5. Cell permeable, highly potent inhibitor of gamma-secretase (IC50 = 0.06 nM).
Catalog No. 50-205-3755