Learn More
Selleck Chemical LLC compound 3i (666-15) 5mg 1433286-70-4
SDP
Supplier:  Selleck Chemical LLC S88465MG
Encompass_Preferred
Compound 3i (666-15) is a potent and selective inhibitor of CREB-mediated gene transcription (IC50 = 0.081 ¿ 0.04 ?M) and also potently inhibits cancer cell growth without harming normal cells. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8387