Learn More
Selleck Chemical LLC Compound 401
SDP
Catalog No. 5000003074
Encompass_Preferred
Compound 401 is a synthetic inhibitor of DNA-PK(IC50 0 28 M) and mTOR (IC50 5 3 M) It has no inhibition on p110 /p85 PI3K ( 100 M) and blocks the phosphorylation of S6 kinase 1 Thr389 and Akt Ser473 in COS7 cells
Catalog No. 50-000-03074 Supplier Selleck Chemical LLC Supplier No. S65065MG
Compound 401 is a synthetic inhibitor of DNA-PK(IC50 0 28 M) and mTOR (IC50 5 3 M) It has no inhibition on p110 /p85 PI3K ( 100 M) and blocks the phosphorylation of S6 kinase 1 Thr389 and Akt Ser473 in COS7 cells