Learn More
Cayman Chemical Compound 401, Purity: ≥98%, CAS: 168425-64-7, 10 mg
SDP
Catalog No. 502985722
Encompass_Preferred
A dual inhibitor of mTOR and DNA-PK (IC50s = 0.28 and 5.3 µM, respectively); selective for DNA-PK and mTOR over PI3K, ATM, and ATR (IC50s = >100 µM for all); inhibits phosphorylation of the mTOR targets S6K1 and Akt in Rat-1 fibroblasts and in M059J glioma cells that lack DNA-PK at 10 µM; inhibits proliferation of TSC1-/- MEFs (IC50 = 2 µM) but not TSC1+/+ MEFs; induces apoptosis in TSC1-/- MEFs
Catalog No. 50-298-5722 Supplier Cayman Chemical Supplier No. 2176910 MG
A dual inhibitor of mTOR and DNA-PK (IC50s = 0.28 and 5.3 µM, respectively); selective for DNA-PK and mTOR over PI3K, ATM, and ATR (IC50s = >100 µM for all); inhibits phosphorylation of the mTOR targets S6K1 and Akt in Rat-1 fibroblasts and in M059J glioma cells that lack DNA-PK at 10 µM; inhibits proliferation of TSC1-/- MEFs (IC50 = 2 µM) but not TSC1+/+ MEFs; induces apoptosis in TSC1-/- MEFs