Learn More
Medchemexpress LLC HY-19341 5mg Medchemexpress, Compound 401 CAS:168425-64-7 Purity:>98%
SDP
Catalog No. 502082120
Encompass_Preferred
Medchemexpress, HY-19341 5mg Compound 401 CAS:168425-64-7 Compound 401 is a synthetic inhibitor of DNA-PK (IC50 = 0.28 μM) that also targets mTOR but not PI3K in vitro. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2120 Supplier Medchemexpress LLC Supplier No. HY193415MG
Medchemexpress, HY-19341 5mg Compound 401 CAS:168425-64-7 Compound 401 is a synthetic inhibitor of DNA-PK (IC50 = 0.28 μM) that also targets mTOR but not PI3K in vitro. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.