Learn More
Abcam Compound 401, DNA-dependent protein kinase and mTOR inhibitor, 10MG
Catalog No. 502707081
Encompass_Preferred
MW 281.31 Da, Purity >99%. Selective, reversible DNA-dependent protein kinase and mTOR inhibitor (IC₅₀ values are 0.28 and 5.3 μM respectively). No affinity for PI3K, ATM and ATR (IC₅₀ values are >100 μM). Able to induce apoptosis in vitro. Cell-permeable.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-7081 Supplier Abcam Supplier No. AB12096810MG
MW 281.31 Da, Purity >99%. Selective, reversible DNA-dependent protein kinase and mTOR inhibitor (IC₅₀ values are 0.28 and 5.3 μM respectively). No affinity for PI3K, ATM and ATR (IC₅₀ values are >100 μM). Able to induce apoptosis in vitro. Cell-permeable.
The product is subject to the following: Abcam Restricted Use Statement