Learn More
Medchemexpress LLC Compound 48/80 10mM 1mL | 94724-12-6 | 519.72 g/mol | C32H45N3O3 | 1 ML
SDP
Catalog No. 5000307889
Encompass_Preferred
Compound 48/80 (poly-p-methoxyphenethylmethylamine) is an oligomeric mast cell activator used to induce mast cell degranulation and histamine release in animal and tissue models. Supplied as a 10 mM solution in DMSO (1 mL), it is intended for research applications studying mast cell signaling and mediator release.
  • Mast cell activator that stimulates G-protein-coupled degranulation pathways.
  • Supplied as a ready-to-use 10 mM solution in DMSO.
  • High reported purity (99.98%) suitable for research use.
  • Applicable to in vitro and in vivo studies of histamine release and signaling.
  • Safety data sheets available for multiple languages.
Catalog No. 50-003-07889 Supplier Medchemexpress LLC Supplier No. HY11576810MM 1ML
Compound 48/80 (poly-p-methoxyphenethylmethylamine) is an oligomeric mast cell activator used to induce mast cell degranulation and histamine release in animal and tissue models. Supplied as a 10 mM solution in DMSO (1 mL), it is intended for research applications studying mast cell signaling and mediator release.
  • Mast cell activator that stimulates G-protein-coupled degranulation pathways.
  • Supplied as a ready-to-use 10 mM solution in DMSO.
  • High reported purity (99.98%) suitable for research use.
  • Applicable to in vitro and in vivo studies of histamine release and signaling.
  • Safety data sheets available for multiple languages.