Learn More
Medchemexpress LLC HY-130592 100mg Medchemexpress, Compound 48/80 (trihydrochloride) CAS:848035-21-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY130592100MG
Encompass_Preferred
Medchemexpress, HY-130592 100mg Compound 48/80 (trihydrochloride) CAS:848035-21-2 Compound 48/80 trihydrochloride (C48/80 trihydrochloride) is a mixture of condensation products of N-methyl-p-methoxyphenethylamine with formaldehyde. Compound 48/80 trihydrochloride is also a histamine releaser and a mast cell degranulator. Compound 48/80 inhibits phosphatidylinositol-specific phospholipase C activity from human platelets . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1245