Learn More
Medchemexpress LLC Compound 48/80 trihydrochloride | 848035-21-2 | 99.8% | 25 MG
SDP
Catalog No. 5000341497
Encompass_Preferred
Compound 48/80 trihydrochloride is a mixture of condensation products of N-methyl-p-methoxyphenethylamine with formaldehyde. It acts as a histamine releaser and a mast cell degranulator, and inhibits phosphatidylinositol-specific phospholipase C activity from human platelets.
  • Acts as a histamine releaser
  • Functions as a mast cell degranulator
  • Inhibits phosphatidylinositol-specific phospholipase C activity
  • Inhibits aggregation of human platelets induced by ADP and PAF-acether
Catalog No. 50-003-41497 Supplier Medchemexpress LLC Supplier No. 25MGHY130592
Compound 48/80 trihydrochloride is a mixture of condensation products of N-methyl-p-methoxyphenethylamine with formaldehyde. It acts as a histamine releaser and a mast cell degranulator, and inhibits phosphatidylinositol-specific phospholipase C activity from human platelets.
  • Acts as a histamine releaser
  • Functions as a mast cell degranulator
  • Inhibits phosphatidylinositol-specific phospholipase C activity
  • Inhibits aggregation of human platelets induced by ADP and PAF-acether