Learn More
Medchemexpress LLC Compound 48/80 trihydrochloride | 848035-21-2 | 99.8% | C32H48Cl3N3O3 | 10MG
SDP
Catalog No. 5000259783
Encompass_Preferred
Compound 48/80 trihydrochloride is an oligomeric mixture formed by condensation of N-methyl-p-methoxyphenethylamine with formaldehyde. It is used in research to trigger mast cell degranulation and histamine release and has been reported to inhibit phosphatidylinositol-specific phospholipase C activity.
  • Used to induce mast cell degranulation and histamine release
  • Oligomeric mixture with molecular formula C32H48Cl3N3O3 and MW 629.10 g·mol⁻¹
  • High supplied purity (approximately 99.8%) suitable for research
  • Available in multiple small mass pack sizes and as a 10 mM aqueous solution
  • Storage: keep solid at 4°C sealed and protect from moisture; in solvent store at -20°C to -80°C for long term
Catalog No. 50-002-59783 Supplier Medchemexpress LLC Supplier No. HY13059210MG
Compound 48/80 trihydrochloride is an oligomeric mixture formed by condensation of N-methyl-p-methoxyphenethylamine with formaldehyde. It is used in research to trigger mast cell degranulation and histamine release and has been reported to inhibit phosphatidylinositol-specific phospholipase C activity.
  • Used to induce mast cell degranulation and histamine release
  • Oligomeric mixture with molecular formula C32H48Cl3N3O3 and MW 629.10 g·mol⁻¹
  • High supplied purity (approximately 99.8%) suitable for research
  • Available in multiple small mass pack sizes and as a 10 mM aqueous solution
  • Storage: keep solid at 4°C sealed and protect from moisture; in solvent store at -20°C to -80°C for long term