Learn More
Cayman Chemical Compound 4880hydroCLOrID 100mg
SDP
Catalog No. 502986660
Encompass_Preferred
A mast cell degranulator; a condensation product of N-methyl-p-methoxyphenethylamine with formaldehyde; supresses CaM (IC50 = 0.41 μg/ml for CaM-dependent Ca2+-transporter ATPase activity) and phospholipase C (IC50s = 2.1 and 5.0 μg/ml for cytosolic and particulate phosphatidylinositol-specific activities, respectively)
Catalog No. 50-298-6660 Supplier Cayman Chemical Supplier No. 22173100 MG
 Restricted Item
Add to Cart
Add to Cart
A mast cell degranulator; a condensation product of N-methyl-p-methoxyphenethylamine with formaldehyde; supresses CaM (IC50 = 0.41 μg/ml for CaM-dependent Ca2+-transporter ATPase activity) and phospholipase C (IC50s = 2.1 and 5.0 μg/ml for cytosolic and particulate phosphatidylinositol-specific activities, respectively)