Learn More
Abcam Compound 56, 5MG
Catalog No. 502681662
Encompass_Preferred
MW 388.3 Da, Purity >97%. Cell-permeable, reversible and ATP-competitive, potent and specific inhibitor of tyrosine kinase activity of EGFR.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-268-1662 Supplier Abcam Supplier No. AB2824435MG
MW 388.3 Da, Purity >97%. Cell-permeable, reversible and ATP-competitive, potent and specific inhibitor of tyrosine kinase activity of EGFR.
The product is subject to the following: Abcam Restricted Use Statement