Learn More
Apexbio Technology LLC Compound 56 171745-13-4 10mM (in 1mL DMSO)
SDP
Catalog No. 5000090327
Encompass_Preferred
Compound 56 (CAS 171745-13-4) known as 4-[(3-bromophenyl)amino]-6 7-diethoxyquinazoline is a highly potent and selective inhibitor of epidermal growth factor receptor (EGFR) tyrosine kinase with an IC50 of 0 006 nM It competitively binds to the ATP-binding site of EGFR blocking EGF-dependent receptor phosphorylation Compound 56 suppresses proliferation and colony formation in human cancer cell lines that overexpress EGFR and inhibits EGF-mediated mitogenesis and oncogenic transformation in fibroblasts with EGFR overexpression It also shows moderate inhibition of HER2/neu tyrosine kinase This compound is widely used to investigate EGFR signaling and function in cancer research
Catalog No. 50-000-90327 Supplier Apexbio Technology LLC Supplier No. A81975.1
Compound 56 (CAS 171745-13-4) known as 4-[(3-bromophenyl)amino]-6 7-diethoxyquinazoline is a highly potent and selective inhibitor of epidermal growth factor receptor (EGFR) tyrosine kinase with an IC50 of 0 006 nM It competitively binds to the ATP-binding site of EGFR blocking EGF-dependent receptor phosphorylation Compound 56 suppresses proliferation and colony formation in human cancer cell lines that overexpress EGFR and inhibits EGF-mediated mitogenesis and oncogenic transformation in fibroblasts with EGFR overexpression It also shows moderate inhibition of HER2/neu tyrosine kinase This compound is widely used to investigate EGFR signaling and function in cancer research