Learn More
Enzo Life Sciences Compound A (25mg). CAS: 14593-25-0
SDP
Supplier:  Enzo Life Sciences ALX550516M025
Encompass_Preferred
Cell permeable, nonsteroidal glucocorticoid receptor modulator with high binding affinity. Exerts anti-inflammatory potential by down-modulating TNF-induced pro-inflammatory gene expression, such as IL-6 and E-selectin. Unlike dexamethasone (BML-EI126) it does not enhance glucocorticoid response element (GRE)-driven gene expression or induce glucocorticoid receptor (GR) binding to GRE-dependent genes in vivo. Alternative name: CpdA, 2-((4-Acetophenyl)-2-chloro-N-methyl)ethylammonium chloride. Purity: ≥98% (HPLC). Solubility: Soluble in DMSO, 100% ethanol or water. Long Term Storage: -80°C.
Catalog No. 50-200-9560