Learn More
TARGETMOL CHEMICALS INC COMPOUND CDY9 5MG
SDP
Catalog No. 502579255
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Compound CDy9 is a highly selective inhibitor of carbonic anhydrase (CA) with an IC50 value of 0.18 uM for hCA II. purity: 99%
Catalog No. 50-257-9255 Supplier TARGETMOL CHEMICALS INC Supplier No. T677745MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Compound CDy9 is a highly selective inhibitor of carbonic anhydrase (CA) with an IC50 value of 0.18 uM for hCA II. purity: 99%