Learn More
AdipoGen Compound E
SDP
Catalog No. 502562370
Encompass_Preferred
Chemical. CAS 209986-17-4. Formula C27H24F2N4O3. MW 490.5. Cell permeable, potent, selective, non-transition state and non-competitive gamma-secretase inhibitor. Notch processing inhibitor. Only weakly affects presenilinase activity.
Catalog No. 50-256-2370 Supplier AdipoGen Supplier No. AGCR10081C250
Chemical. CAS 209986-17-4. Formula C27H24F2N4O3. MW 490.5. Cell permeable, potent, selective, non-transition state and non-competitive gamma-secretase inhibitor. Notch processing inhibitor. Only weakly affects presenilinase activity.