Learn More
Medchemexpress LLC Compound E | 209986-17-4 | 99.97% | C27H24F2N4O3 | 1 ML
SDP
Catalog No. 5000356541
Encompass_Preferred
Compound E is a γ-secretase inhibitor. It blocks β-amyloid(40), β-amyloid(42), and Notch γ-secretase cleavage, making it suitable for various research applications.
  • Blocks β-amyloid(40), β-amyloid(42), and Notch γ-secretase cleavage
  • Inhibits gamma secretase-mediated amyloid beta (1 to 40) production
  • Reduces proliferation of T47D and MCF-7 cell lines
Catalog No. 50-003-56541 Supplier Medchemexpress LLC Supplier No. HY1417610MM/1ML
Compound E is a γ-secretase inhibitor. It blocks β-amyloid(40), β-amyloid(42), and Notch γ-secretase cleavage, making it suitable for various research applications.
  • Blocks β-amyloid(40), β-amyloid(42), and Notch γ-secretase cleavage
  • Inhibits gamma secretase-mediated amyloid beta (1 to 40) production
  • Reduces proliferation of T47D and MCF-7 cell lines