Learn More
Abcam Compound E, gamma;-secretase inhibitor, 250UG
Catalog No. NC1609905
Encompass_Preferred
MW 490.5 Da, Purity >99%. Cell-permeable, selective, non-competitive, potent γ-secretase inhibitor (IC₅₀ = 0.3 nM). Active in vitro and in vivo.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC1609905 Supplier Abcam Supplier No. AB142164250UG
MW 490.5 Da, Purity >99%. Cell-permeable, selective, non-competitive, potent γ-secretase inhibitor (IC₅₀ = 0.3 nM). Active in vitro and in vivo.
The product is subject to the following: Abcam Restricted Use Statement