Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Compound E-3P-MDM2 | 99.8% | 1463.55 g/mol | C77H93Cl2N13O12 | 10 MG
SDP

Catalog No. 5000288267
Encompass_Preferred

PROTAC EZH2 Degrader-2 is a proteolysis-targeting chimera (PROTAC) that induces dose-dependent degradation of EZH2 and shows anti-proliferative and pro-apoptotic activity in cell models. It is supplied as a high-purity solid for research use.

  • High purity (99.78%).
  • White to off-white solid appearance.
  • Molecular weight approximately 1,463.55 g/mol.
  • Soluble in DMSO at 100 mg/mL; may require ultrasonic agitation.
  • Stability: store solids at 4°C under nitrogen; in solution, -80°C (6 months) or -20°C (1 month).
  • Intended for biochemical and cell-based studies of EZH2 degradation and related anticancer research.

Catalog No. 50-002-88267 Supplier Medchemexpress LLC Supplier No. HY15716410MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

PROTAC EZH2 Degrader-2 is a proteolysis-targeting chimera (PROTAC) that induces dose-dependent degradation of EZH2 and shows anti-proliferative and pro-apoptotic activity in cell models. It is supplied as a high-purity solid for research use.

  • High purity (99.78%).
  • White to off-white solid appearance.
  • Molecular weight approximately 1,463.55 g/mol.
  • Soluble in DMSO at 100 mg/mL; may require ultrasonic agitation.
  • Stability: store solids at 4°C under nitrogen; in solution, -80°C (6 months) or -20°C (1 month).
  • Intended for biochemical and cell-based studies of EZH2 degradation and related anticancer research.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.