Learn More
AdipoGen Compound E (5 mg)
SDP
Supplier:  AdipoGen AGCR10081M005
Encompass_Preferred
Chemical. CAS: 209986-17-4. Formula: C27H24F2N4O3. MW: 490.5. Cell permeable, potent, selective, non-transition state and non-competitive gamma-secretase inhibitor. Notch processing inhibitor. Only weakly affects presenilinase activity.
Catalog No. 50-205-3826