Learn More
Abcam Compound E, gamma;-secretase inhibitor, 1MG
Catalog No. 502583323
Encompass_Preferred
MW 490.5 Da, Purity >99%. Cell-permeable, selective, non-competitive, potent γ-secretase inhibitor (IC₅₀ = 0.3 nM). Active in vitro and in vivo.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-258-3323 Supplier Abcam Supplier No. AB1421641MG
MW 490.5 Da, Purity >99%. Cell-permeable, selective, non-competitive, potent γ-secretase inhibitor (IC₅₀ = 0.3 nM). Active in vitro and in vivo.
The product is subject to the following: Abcam Restricted Use Statement