Learn More
STEMCELL Technologies Compound E, Size: 500 µg
SDP
Supplier:  STEMCELL Technologies 73952
Encompass
Compound E is a potent, cell-permeable, selective inhibitor that blocks the cleavage of both gamma-secretase and the Notch intracellular domain with similar IC50 values, ranging from 0.24 to 0.37 nM (Beher et al.; Seiffert et al.). DIFFERENTIATION· Inhibits growth, differentiation, and motility of neuroblastoma cells (Ferrari-Toninelli et al.).· Accelerates the differentiation of human embryonic stem cells into primitive neural stem cells (Li et al.).
Catalog No. 50-197-6154
Add to Cart
Description
Description
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov