Learn More
Enzo Life Sciences Compound E, (250µg), CAS Number: 209986-17-4
SDP
Catalog No. 501031739
Encompass_Preferred
γ-Secretase inhibitor. Cell permeable, potent, selective, non-transition state and non-competitive inhibitor of γ-secretase (IC50=0.3nM for total β-amyloid) and Notch processing, which ceases cell differentiation. At higher concentrations (20-400µM), only weakly affects the presenilase activity. Activity tested!. Alternative Name: (2S)-2-{[(3,5-Difluorophenyl)acetyl]amino}-N-[(3S)-1-methyl-2-oxo-5-phenyl-2,3-dihydro-1H-1,4-benzodiazepin-3-yl]propanamide, γ-Secretase Inhibitor, GSI. Formula: C27H24F2N4O3. MW: 490.5. CAS Number: 209986-17-4. Purity: ≥95% (HPLC). Appearance: White to off-white powder. Solubility: Soluble in DMSO. Long Term Storage: -20°C
Catalog No. 50-103-1739 Supplier Enzo Life Sciences Supplier No. ALX270415C250
γ-Secretase inhibitor. Cell permeable, potent, selective, non-transition state and non-competitive inhibitor of γ-secretase (IC50=0.3nM for total β-amyloid) and Notch processing, which ceases cell differentiation. At higher concentrations (20-400µM), only weakly affects the presenilase activity. Activity tested!. Alternative Name: (2S)-2-{[(3,5-Difluorophenyl)acetyl]amino}-N-[(3S)-1-methyl-2-oxo-5-phenyl-2,3-dihydro-1H-1,4-benzodiazepin-3-yl]propanamide, γ-Secretase Inhibitor, GSI. Formula: C27H24F2N4O3. MW: 490.5. CAS Number: 209986-17-4. Purity: ≥95% (HPLC). Appearance: White to off-white powder. Solubility: Soluble in DMSO. Long Term Storage: -20°C