Learn More
Cayman Chemical Comprehensive Fatty Acid Methyl Ester Mixture 3, 1 mL
SDP
Catalog No. 502990199
Encompass_Preferred
A mixture of FAMEs; intended for use as a reference standard for the quantification of these fatty acids in various sample types by GC-FID or GC-MS
Catalog No. 50-299-0199 Supplier Cayman Chemical Supplier No. 293711 ML
A mixture of FAMEs; intended for use as a reference standard for the quantification of these fatty acids in various sample types by GC-FID or GC-MS