Learn More
Cayman Chemical Comprehensive Fatty Acid Methyl Ester Mixture 2, 1 mL
SDP
Catalog No. 502989147
Encompass_Preferred
A mixture of FAMEs; intended for use as a reference standard for the quantification of these fatty acids in various sample types by GC-FID or GC-MS
Catalog No. 50-298-9147 Supplier Cayman Chemical Supplier No. 293351 ML
A mixture of FAMEs; intended for use as a reference standard for the quantification of these fatty acids in various sample types by GC-FID or GC-MS