Learn More
Cayman Chemical compre KInase ScreenIn 50uL
SDP
Catalog No. 502941611
Encompass_Preferred
A mixture of farnesene isomers may contain trans-b-farnesene cis-a-farnesene trans-a-farnesene and bisabolene among others
Catalog No. 50-294-1611 Supplier Cayman Chemical Supplier No. 2894050 UL
Add to Cart
Add to Cart
Request a Quote
A mixture of farnesene isomers may contain trans-b-farnesene cis-a-farnesene trans-a-farnesene and bisabolene among others