Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Microbiologics Comprehensive Human Papillomavirus (HPV) Control Panel
SDP

Catalog No. 21402130 Shop All Microbiologics Products
Change view
Click to view available options
Level:
1
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Level
21-402-130 1
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 21-402-130 Supplier Microbiologics Supplier No. 8257
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The Comprehensive Human Papillomavirus (HPV) Control Panel consists of 18 individually packaged lyophilized pellets: six pellets of positive control 1, six pellets of positive control 2, and six pellets of positive control 3.

Positive Control 1:

  • HPV Type 31 Surrogate, gene target E6/E7
  • HPV Type 31 Surrogate, gene target L1
  • HPV Type 33 Surrogate, gene target E6/E7
  • HPV Type 66 Surrogate, gene target E6/E7
  • HPV Type 68 Surrogate, gene target E6/E7
  • HPV Type 68 Surrogate, gene target L1
  • Human Cells

Positive Control 2:

  • HPV Type 16 Surrogate, gene target E6/E7
  • HPV Type 16 Surrogate, gene target L1
  • HPV Type 18 Surrogate, gene target E6/E7
  • HPV Type 18 Surrogate, gene target L1
  • HPV Type 35 Surrogate, gene target E6/E7
  • HPV Type 45 Surrogate, gene target E6/E7
  • HPV Type 45 Surrogate, gene target L1
  • HPV Type 59 Surrogate, gene target E6/E7
  • Human Cells

Positive Control 3:

  • HPV Type 39 Surrogate, gene target E6/E7
  • HPV Type 51 Surrogate, gene target E6/E7
  • HPV Type 52 Surrogate, gene target E6/E7
  • HPV Type 56 Surrogate, gene target E6/E7
  • HPV Type 58 Surrogate, gene target E6/E7
  • Human Cells

Specifications

Sample Type Human Patient Samples
Type Control Panel
Disposable Single-use
Level 1
Sterility Non-sterile
Form Pellet
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.