Learn More
Chemglass Life Sciences Chem-Thread Compression Cap with Aperture, Size #9, Accepts 7-9mm OD Tubing
SDP
Catalog No. CG35202
Encompass_Preferred
Chem-Thread Compression Cap with Aperture, Size #9, Accepts 7-9mm OD Tubing. Celcon compression cap with listed hole size for Chem-Threads.
Catalog No. CG35202 Supplier Chemglass Life Sciences Supplier No. CG35202
Chem-Thread Compression Cap with Aperture, Size #9, Accepts 7-9mm OD Tubing. Celcon compression cap with listed hole size for Chem-Threads.