Learn More
METTLER TOLEDO COMPRESSION FITTING 1/2 NPT
Catalog No. 502741724 Shop All METTLER TOLEDO Products
Encompass
Compression fitting 1/2" NPT 6mm bore
Catalog No. 50-274-1724 Supplier METTLER TOLEDO Supplier No. 30245645
Compression fitting 1/2" NPT 6mm bore