Learn More
Bellco Glass, Inc. Compression Fitting, 12mm For 18mm Port
SDP
Catalog No. 501946440
Encompass_Preferred
Bellco compression fittings are designed using our standard 45mm blue polypropylene open top caps and white silicone gaskets. A wide variety of fittings are available to allow pH and OD monitoring via insertion of these probes into sidearms or vessels. Bellco probe adapters are designed to allow introduction of threaded probes into vessels through 200mm stainless steel or a 45mm vertical sidearm.. MADE IN THE USA
Catalog No. 50-194-6440 Supplier Bellco Glass, Inc. Supplier No. 790900030
Bellco compression fittings are designed using our standard 45mm blue polypropylene open top caps and white silicone gaskets. A wide variety of fittings are available to allow pH and OD monitoring via insertion of these probes into sidearms or vessels. Bellco probe adapters are designed to allow introduction of threaded probes into vessels through 200mm stainless steel or a 45mm vertical sidearm.. MADE IN THE USA