Learn More
Chemglass Life Sciences 14mm PTFE Compression Fitting for Lasentec Probe, 3/4" Male NPT
SDP
Catalog No. 501531280
Encompass_Preferred
14mm PTFE Compression Fitting for Lasentec Probe, 3/4" Male NPT. 14mm PTFE compression fitting for 10 X 10 compound angle side neck. Component of CG-1945-T-50 PTFE Reaction Vessel Lid. Image shown is the fully assembled reaction vessel lid. Please note that this item is only the compression fitting.
Catalog No. 50-153-1280 Supplier Chemglass Life Sciences Supplier No. CG1945T102
14mm PTFE Compression Fitting for Lasentec Probe, 3/4" Male NPT. 14mm PTFE compression fitting for 10 X 10 compound angle side neck. Component of CG-1945-T-50 PTFE Reaction Vessel Lid. Image shown is the fully assembled reaction vessel lid. Please note that this item is only the compression fitting.