Learn More
Chemglass Life Sciences 19mm PTFE Compression Fitting for Lasentec Probe, 3/4" Male NPT
SDP
Catalog No. 501531281
Encompass_Preferred
19mm PTFE Compression Fitting for Lasentec Probe, 3/4" Male NPT. Component of CG-1945-T-50 PTFE Reaction Vessel Lid. Image shown is the fully assembled reaction vessel lid. Please note that this item is only the compression fitting.
Catalog No. 50-153-1281 Supplier Chemglass Life Sciences Supplier No. CG1945T106
19mm PTFE Compression Fitting for Lasentec Probe, 3/4" Male NPT. Component of CG-1945-T-50 PTFE Reaction Vessel Lid. Image shown is the fully assembled reaction vessel lid. Please note that this item is only the compression fitting.