Learn More
AVANTOR MASTERFLEX (VWR LLC) Fitting, PFA, Straight, Compression Union, 4mm OD
Catalog No. NC9460169
Encompass
Able to withstand higher pressures than barbed fittings, compression fittings are an excellent choice to connect rigid tubing systems.
Catalog No. NC9460169 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX3132161
Able to withstand higher pressures than barbed fittings, compression fittings are an excellent choice to connect rigid tubing systems.