Learn More
Bellco Glass, Inc. Compression Fitting Assembly , 1/4
SDP
Catalog No. 501946427
Encompass_Preferred
316L stainless steel compression fitting for use with 1/4" tubes. Useful for addition and withdrawal of contents from bioreactor vessel. Can fit 10mm port on 3L and 7L metric bioreactors.. MADE IN THE USA
Catalog No. 50-194-6427 Supplier Bellco Glass, Inc. Supplier No. 790900014
316L stainless steel compression fitting for use with 1/4" tubes. Useful for addition and withdrawal of contents from bioreactor vessel. Can fit 10mm port on 3L and 7L metric bioreactors.. MADE IN THE USA