Learn More
Bellco Glass, Inc. Compression Fitting Assembly , 1/8 For 10mm Port
SDP
Catalog No. 501946429
Encompass_Preferred
Bellco compression fittings are designed using our standard 45mm blue polypropylene open top caps and white silicone gaskets. A wide variety of fittings are available to allow pH and OD monitoring via insertion of these probes into sidearms or vessels. Bellco probe adapters are designed to allow introduction of threaded probes into vessels through 200mm stainless steel or a 45mm vertical sidearm.. MADE IN THE USA
Catalog No. 50-194-6429 Supplier Bellco Glass, Inc. Supplier No. 790900018
Bellco compression fittings are designed using our standard 45mm blue polypropylene open top caps and white silicone gaskets. A wide variety of fittings are available to allow pH and OD monitoring via insertion of these probes into sidearms or vessels. Bellco probe adapters are designed to allow introduction of threaded probes into vessels through 200mm stainless steel or a 45mm vertical sidearm.. MADE IN THE USA