Learn More
Chemglass Life Sciences 1/2" PFA Tee Compression Fitting
SDP
Catalog No. 50143587
Encompass_Preferred
1/2" PFA Tee Compression Fitting. PFA Tee compression fittings are ideally suited for use in corrosive environments and chemical applications. The fitting design yields a leak-tight connection and requires no special tools to assemble. The connection is achieved by merely inserting the tubing into the fitting assembly and tightening the nut.
Catalog No. 50-143-587 Supplier Chemglass Life Sciences Supplier No. CG1975T22
1/2" PFA Tee Compression Fitting. PFA Tee compression fittings are ideally suited for use in corrosive environments and chemical applications. The fitting design yields a leak-tight connection and requires no special tools to assemble. The connection is achieved by merely inserting the tubing into the fitting assembly and tightening the nut.