Learn More
Chemglass Life Sciences 1/4" to 1/4" PFA Compression Fitting
SDP
Catalog No. 50143611
Encompass_Preferred
1/4" to 1/4" PFA Compression Fitting. Designed for use with our CG-1975 heavy wall PTFE tubing. Fittings are made entirely of chemically inert corrosion resistant PFA PTFE. Sizes and configurations other than those listed are available.
Catalog No. 50-143-611 Supplier Chemglass Life Sciences Supplier No. CG197519
1/4" to 1/4" PFA Compression Fitting. Designed for use with our CG-1975 heavy wall PTFE tubing. Fittings are made entirely of chemically inert corrosion resistant PFA PTFE. Sizes and configurations other than those listed are available.