Learn More
Chemglass Life Sciences 1/8" to 1/2" PFA Compression Fitting
SDP
Catalog No. 50143648
Encompass_Preferred
1/8" to 1/2" PFA Compression Fitting. Designed for use with our CG-1975 heavy wall PTFE tubing. Fittings are made entirely of chemically inert corrosion resistant PFA PTFE. Sizes and configurations other than those listed are available.
Catalog No. 50-143-648 Supplier Chemglass Life Sciences Supplier No. CG197518
1/8" to 1/2" PFA Compression Fitting. Designed for use with our CG-1975 heavy wall PTFE tubing. Fittings are made entirely of chemically inert corrosion resistant PFA PTFE. Sizes and configurations other than those listed are available.